ENA321763447 Enamine

methyl 2-chloroquinoline-3-carboxylate

  • MDL number MFCD06203870
  • Cas Number : 16498-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75167

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75167
Availability details for
Quantity
  • Availability
  • Add to Cart