ENA321763452 Enamine

5-fluoro-N-(oxan-4-yl)pyridin-2-amine

  • MDL number MFCD14621370
  • Cas Number : 1248980-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75172

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75172
Availability details for
Quantity
  • Availability
  • Add to Cart