ENA321763453 Enamine

2-(6-amino-2,3-dihydro-1H-indol-1-yl)-N-methylacetamide

  • MDL number MFCD11137672
  • Cas Number : 1019555-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74460
Availability details for
Quantity
  • Availability
  • Add to Cart