ENA321763454 Enamine

5-(4-cyclopropanecarbonylpiperazin-1-yl)pyridin-2-amine

  • MDL number MFCD11982080
  • Cas Number : 1154964-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75176
Availability details for
Quantity
  • Availability
  • Add to Cart