ENA321763456 Enamine

2-chloro-6,7-difluoroquinoxaline

  • MDL number MFCD11518958
  • Cas Number : 143007-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75178
Availability details for
Quantity
  • Availability
  • Add to Cart