ENA321763459 Enamine

3-(chloromethyl)-5H,6H,7H-[1,3]thiazolo[3,2-a]pyrimidine hydrochloride

  • MDL number MFCD18785528
  • Cas Number : 62835-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75183
Availability details for
Quantity
  • Availability
  • Add to Cart