ENA321763464 Enamine

2-[(3-chlorophenyl)amino]acetonitrile

  • MDL number MFCD11152884
  • Cas Number : 24889-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75189
Availability details for
Quantity
  • Availability
  • Add to Cart