ENA321763468 Enamine

4-(3,5-dimethyl-1H-pyrazol-1-yl)-3-fluorobenzoic acid

  • MDL number MFCD11156568
  • Cas Number : 1152868-20-6
  • Molecular Weight : 234.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75198
Availability details for
Quantity
  • Availability
  • Add to Cart