ENA321763469 Enamine

methyl 2-amino-5-tert-butylthiophene-3-carboxylate

  • MDL number MFCD08059073
  • Cas Number : 216574-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75199

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75199
Availability details for
Quantity
  • Availability
  • Add to Cart