ENA321763470 Enamine

1-(2-chlorophenyl)-3-(methylamino)pyrrolidin-2-one

  • MDL number MFCD14689790
  • Cas Number : 1248430-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74463
Availability details for
Quantity
  • Availability
  • Add to Cart