ENA321763471 Enamine

3-chloro-6-(propan-2-yl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD08444917
  • Cas Number : 923759-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75202
Availability details for
Quantity
  • Availability
  • Add to Cart