ENA321763473 Enamine

4-phenylbenzene-1-sulfonamide

  • MDL number MFCD03731839
  • Cas Number : 4371-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75204

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75204
Availability details for
Quantity
  • Availability
  • Add to Cart