ENA321763477 Enamine

(1-methanesulfonylpropan-2-yl)(methyl)amine

  • MDL number MFCD16860227
  • Cas Number : 1248451-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75209
Availability details for
Quantity
  • Availability
  • Add to Cart