ENA321763480 Enamine

2-phenoxybutan-1-amine

  • MDL number MFCD16081586
  • Cas Number : 6440-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75212
Availability details for
Quantity
  • Availability
  • Add to Cart