ENA321763485 Enamine

[3-(2,2,2-trifluoroethoxy)phenyl]methanol

  • MDL number MFCD11149514
  • Cas Number : 1021063-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74466
Availability details for
Quantity
  • Availability
  • Add to Cart