ENA321763491 Enamine

2-(2-ethyl-1H-1,3-benzodiazol-1-yl)acetonitrile

  • MDL number MFCD05722947
  • Cas Number : 54980-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75226

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75226
Availability details for
Quantity
  • Availability
  • Add to Cart