ENA321763493 Enamine

2-methyl-1-(2-methylphenyl)propan-1-amine

  • MDL number MFCD12154056
  • Cas Number : 860701-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75228
Availability details for
Quantity
  • Availability
  • Add to Cart