ENA321763499 Enamine

1-(4-amino-2-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD16074941
  • Cas Number : 853910-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75236
Availability details for
Quantity
  • Availability
  • Add to Cart