ENA321763503 Enamine

3-(2,5-difluorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD11639661
  • Cas Number : 1096348-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75240
Availability details for
Quantity
  • Availability
  • Add to Cart