ENA321763506 Enamine

4-(2-bromophenyl)-1H-pyrazol-5-amine

  • MDL number MFCD11168884
  • Cas Number : 301373-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75247
Availability details for
Quantity
  • Availability
  • Add to Cart