ENA321763512 Enamine

1-(1-trifluoromethanesulfonylpiperidin-4-yl)methanamine

  • MDL number MFCD12086206
  • Cas Number : 1154673-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75254
Availability details for
Quantity
  • Availability
  • Add to Cart