ENA321763513 Enamine

2-[(3-bromophenyl)amino]-N-(2-methylpropyl)propanamide

  • MDL number MFCD11176594
  • Cas Number : 1132788-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74484
Availability details for
Quantity
  • Availability
  • Add to Cart