ENA321763517 Enamine

N-ethylpyridin-4-amine

  • MDL number MFCD00082661
  • Cas Number : 35036-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75265
Availability details for
Quantity
  • Availability
  • Add to Cart