ENA321763518 Enamine

2-(piperidin-3-yl)-1-(propan-2-yl)-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD11506440
  • Cas Number : 1185300-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75266
Availability details for
Quantity
  • Availability
  • Add to Cart