ENA321763527 Enamine

5-(3-methoxyphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00628650
  • Cas Number : 247109-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75276
Availability details for
Quantity
  • Availability
  • Add to Cart