ENA321763528 Enamine

pyrrolidine-3-sulfonamide

  • MDL number MFCD12913289
  • Cas Number : 1208507-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75277
Availability details for
Quantity
  • Availability
  • Add to Cart