ENA321763530 Enamine

N-methyl-4-(methylamino)pyridine-2-carboxamide

  • MDL number MFCD11504981
  • Cas Number : 1065074-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75280
Availability details for
Quantity
  • Availability
  • Add to Cart