ENA321763532 Enamine

N-ethyl-6-methylpyridazin-3-amine

  • MDL number MFCD14621076
  • Cas Number : 1250628-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75283
Availability details for
Quantity
  • Availability
  • Add to Cart