ENA321763539 Enamine

[(4-fluorophenyl)(pyrazin-2-yl)methyl](methyl)amine

  • MDL number MFCD12154775
  • Cas Number : 1184512-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75290
Availability details for
Quantity
  • Availability
  • Add to Cart