ENA321763541 Enamine

2-(4-bromophenoxy)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD01409051
  • Cas Number : 1181448-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75293
Availability details for
Quantity
  • Availability
  • Add to Cart