ENA321763546 Enamine

3-oxo-3-(thiomorpholin-4-yl)propanenitrile

  • MDL number MFCD00276561
  • Cas Number : 15029-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75312
Availability details for
Quantity
  • Availability
  • Add to Cart