ENA321763548 Enamine

octahydropyrrolo[1,2-a]pyrazin-1-one

  • MDL number MFCD00601658
  • Cas Number : 16620-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74500
Availability details for
Quantity
  • Availability
  • Add to Cart