ENA321763557 Enamine

3-bromo-4-methoxyaniline

  • MDL number MFCD04094339
  • Cas Number : 19056-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75325

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-75325
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart