ENA321763563 Enamine

2-(phenylamino)butanamide

  • MDL number MFCD12817083
  • Cas Number : 1218556-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75332
Availability details for
Quantity
  • Availability
  • Add to Cart