ENA321763567 Enamine

1-amino-2-cyclopropylpropan-2-ol

  • MDL number MFCD12169410
  • Cas Number : 868851-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75338

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75338
Availability details for
Quantity
  • Availability
  • Add to Cart