ENA321763569 Enamine

2-(pentylamino)ethan-1-ol

  • MDL number MFCD00014031
  • Cas Number : 35161-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75340
Availability details for
Quantity
  • Availability
  • Add to Cart