ENA321763571 Enamine

4-oxo-1,4-dihydrocinnoline-3-carboxylic acid

  • MDL number MFCD18483294
  • Cas Number : 18514-85-7
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74333
Availability details for
Quantity
  • Availability
  • Add to Cart