ENA321763572 Enamine

3-amino-1-(2-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD14689574
  • Cas Number : 1247157-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75347
Availability details for
Quantity
  • Availability
  • Add to Cart