ENA321763573 Enamine

5-[(3-fluoro-4-methoxyphenyl)methyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD11207464
  • Cas Number : 1094350-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75348
Availability details for
Quantity
  • Availability
  • Add to Cart