ENA321763576 Enamine

N-(4-chlorophenyl)-2-(propylamino)acetamide

  • MDL number MFCD11154493
  • Cas Number : 1021230-54-5
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75352
Availability details for
Quantity
  • Availability
  • Add to Cart