ENA321763578 Enamine

(2-aminoethyl)(butyl)methylamine

  • MDL number MFCD00047952
  • Cas Number : 19435-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75354
Availability details for
Quantity
  • Availability
  • Add to Cart