ENA321763581 Enamine

2-(2-methanesulfonamidophenyl)acetic acid

  • MDL number MFCD11052342
  • Cas Number : 1047724-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74508
Availability details for
Quantity
  • Availability
  • Add to Cart