ENA321763582 Enamine

1-(2-fluorophenoxy)-3,3-dimethylbutan-2-amine

  • MDL number MFCD09738172
  • Cas Number : 954272-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75358
Availability details for
Quantity
  • Availability
  • Add to Cart