ENA321763585 Enamine

1-amino-3,3-dimethylbutan-2-ol

  • MDL number MFCD09260550
  • Cas Number : 70489-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-57
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75363
Availability details for
Quantity
  • Availability
  • Add to Cart