ENA321763586 Enamine

2-(1-hydroxy-2-methylpropan-2-yl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD01646565
  • Cas Number : 4490-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74509
Availability details for
Quantity
  • Availability
  • Add to Cart