ENA321763587 Enamine

2-(2-aminoethyl)-5-methoxyphenol

  • MDL number MFCD08448782
  • Cas Number : 54410-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75366
Availability details for
Quantity
  • Availability
  • Add to Cart