ENA321763588 Enamine

4-iodo-N-methylbenzene-1-sulfonamide

  • MDL number MFCD11647536
  • Cas Number : 134283-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75367
Availability details for
Quantity
  • Availability
  • Add to Cart