ENA321763589 Enamine

N-ethyl-4-iodobenzene-1-sulfonamide

  • MDL number MFCD07359355
  • Cas Number : 881302-29-0
  • Molecular Weight : 311.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75368
Availability details for
Quantity
  • Availability
  • Add to Cart