ENA321763591 Enamine

N-(2-aminoethyl)-3-hydroxypyridine-2-carboxamide

  • MDL number MFCD12772813
  • Cas Number : 1281694-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74334
Availability details for
Quantity
  • Availability
  • Add to Cart