ENA321763593 Enamine

N-(2-iodophenyl)methanesulfonamide

  • MDL number MFCD00978524
  • Cas Number : 116547-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75372
Availability details for
Quantity
  • Availability
  • Add to Cart